Please login to your online account to display your discounted pricing

ethyle3-bromobenzoate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC308541000

 View more versions of this product

Catalog No. AC308541000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 229.08
Physical Form Liquid
Quantity 100g
Color Clear colorless
Density 1.43g/mL
Linear Formula BrC6H4COCH5
Molecular Formula C9H9BrO2
Assay Percent Range 98%
CAS 24398-88-7
Chemical Name or Material ethyle3-bromobenzoate
MDL Number MFCD00013529
Packaging Glass bottle
Refractive Index 1.5405 to 1.5425 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material ethyle3-bromobenzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9BrO2
Linear Formula BrC6H4COCH5
Formula Weight 229.08
MDL Number MFCD00013529
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents