Learn More
Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 118000500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 3-aminobenzoate, methanesulfonic acid salt | |
146.0°C to 150.0°C | |
Authentic | |
Plastic bottle | |
H2NC6H4CO2C2H5·CH3SO3H | |
50 g | |
15, 9786 | |
(5% in Water) Clear solution | |
CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O | |
261.29 | |
261.29 | |
Fine Powder |
886-86-2 | |
White | |
98% | |
C9H11NO2·CH4O3S | |
MFCD00013176 | |
14, 389 | |
ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt | |
FQZJYWMRQDKBQN-UHFFFAOYSA-N | |
ethyl 3-aminobenzoate;methanesulfonic acid | |
261501 | |
98% |
Chemical Identifiers
886-86-2 | |
261.29 | |
FQZJYWMRQDKBQN-UHFFFAOYSA-N | |
261501 | |
CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O |
C9H11NO2·CH4O3S | |
MFCD00013176 | |
ethyl 3-aminobenzoate methanesulfonate, tricaine, tricaine methanesulfonate, finquel, metacaine, tricaine mesylate, 3-aminobenzoic acid ethyl ester methanesulfonate, ethyl m-aminobenzoate methanesulfonate, ethyl 3-aminobenzoate methanesulfonic acid salt, 3-aminobenzoic acid ethyl ester methanesulfonate salt | |
ethyl 3-aminobenzoate;methanesulfonic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 212-956-8
RUO – Research Use Only