Please login to your online account to display your discounted pricing

ethyle3,5-dichlorobenzoate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL00759EA

 View more versions of this product

Catalog No. TL00759EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 219.07
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C9H8Cl2O2
Linear Formula Cl2C6H3C(O)OCH2CH3
Chemical Name or Material Ethyl 3, 5-dichlorobenzoate
MDL Number MFCD00221467
CAS 91085-56-2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ethyl 3, 5-dichlorobenzoate
CAS 91085-56-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8Cl2O2
Linear Formula Cl2C6H3C(O)OCH2CH3
Formula Weight 219.07
MDL Number MFCD00221467
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents