Please login to your online account to display your discounted pricing

ethyle3-(4-hydroxyphenyl)propanoate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB00914EA

 View more versions of this product

Catalog No. BTB00914EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 194.23
Quantity 10g
Assay 97%
CAS Max % 100.0
Molecular Formula C11H14O3
Linear Formula CH3CH2OC(O)(CH2)2C6H4OH
MDL Number MFCD00020209
CAS 23795-02-0
Assay Percent Range 97%
CAS Min % 96.5
Chemical Name or Material ethyle3-(4-hydroxyphenyl)propanoate
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material ethyle3-(4-hydroxyphenyl)propanoate
CAS 23795-02-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H14O3
Linear Formula CH3CH2OC(O)(CH2)2C6H4OH
Formula Weight 194.23
MDL Number MFCD00020209