Please login to your online account to display your discounted pricing

ethyle3-(4-hydroxyphenyl)propanoate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB00914DA

 View more versions of this product

Catalog No. BTB00914DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 194.23
Quantity 1g
Assay 97%
CAS Max % 100.0
Linear Formula CH3CH2OC(O)(CH2)2C6H4OH
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00020209
Chemical Name or Material ethyle3-(4-hydroxyphenyl)propanoate
CAS Min % 96.5
Molecular Formula C11H14O3
CAS 23795-02-0

Specifications

Chemical Name or Material ethyle3-(4-hydroxyphenyl)propanoate
CAS 23795-02-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H14O3
Linear Formula CH3CH2OC(O)(CH2)2C6H4OH
Formula Weight 194.23
MDL Number MFCD00020209