missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ethyle3-(4-hydroxyphenyl)propanoate, 97%, Maybridge™
$19.98 - $19.98
Chemical Identifiers
CAS | 23795-02-0 |
---|---|
Molecular Formula | C11H14O3 |
Molecular Weight (g/mol) | 194.23 |
MDL Number | MFCD00020209 |
InChI Key | UFMFPPAZUJDUMY-UHFFFAOYSA-N |
Synonym | benzenepropanoic acid, 4-hydroxy-, ethyl ester, ethyl 3-4-hydroxyphenyl propanoate, ethyl 3-4-hydroxyphenyl propionate, ethyl 4-hydroxybenzenepropanoate, ethyl 4-hydroxyhydrocinnamate, ethyl 4-hydroxyphenyl-3-propanoate, ethyl p-hydroxyhydrocinnamate, ethyl phloretate, ethyl4-hydroxyhydrocinnamate, pubchem20092 |
PubChem CID | 2736435 |
IUPAC Name | ethyl 3-(4-hydroxyphenyl)propanoate |
SMILES | CCOC(=O)CCC1=CC=C(C=C1)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BTB00914DA
|
Fisher Scientific Company
BTB00914DA |
1g | Amber Glass Bottle |
Each for $19.98
|
|
BTB00914EA
|
Fisher Scientific Company
BTB00914EA |
10g | Amber Glass Bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB00914EB
|
Fisher Scientific Company
BTB00914EB |
25g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
23795-02-0 | |
194.23 | |
UFMFPPAZUJDUMY-UHFFFAOYSA-N | |
2736435 | |
CCOC(=O)CCC1=CC=C(C=C1)O |
C11H14O3 | |
MFCD00020209 | |
benzenepropanoic acid, 4-hydroxy-, ethyl ester, ethyl 3-4-hydroxyphenyl propanoate, ethyl 3-4-hydroxyphenyl propionate, ethyl 4-hydroxybenzenepropanoate, ethyl 4-hydroxyhydrocinnamate, ethyl 4-hydroxyphenyl-3-propanoate, ethyl p-hydroxyhydrocinnamate, ethyl phloretate, ethyl4-hydroxyhydrocinnamate, pubchem20092 | |
ethyl 3-(4-hydroxyphenyl)propanoate |
Specifications
Amber Glass Bottle | |
23795-02-0 | |
100.0 | |
C11H14O3 | |
MFCD00020209 | |
UFMFPPAZUJDUMY-UHFFFAOYSA-N | |
ethyl 3-(4-hydroxyphenyl)propanoate | |
2736435 | |
97% |
1g | |
96.5 | |
97% | |
CH3CH2OC(O)(CH2)2C6H4OH | |
benzenepropanoic acid, 4-hydroxy-, ethyl ester, ethyl 3-4-hydroxyphenyl propanoate, ethyl 3-4-hydroxyphenyl propionate, ethyl 4-hydroxybenzenepropanoate, ethyl 4-hydroxyhydrocinnamate, ethyl 4-hydroxyphenyl-3-propanoate, ethyl p-hydroxyhydrocinnamate, ethyl phloretate, ethyl4-hydroxyhydrocinnamate, pubchem20092 | |
CCOC(=O)CCC1=CC=C(C=C1)O | |
194.23 | |
194.23 | |
ethyle3-(4-hydroxyphenyl)propanoate |