missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle3-(4-hydroxyphenyl)propanoate, 97%, Maybridge

Manufacturer:  Fisher Scientific Company BTB00914EB

 View more versions of this product

Catalog No. BTB00914EB

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

194.23
97%
CH3CH2OC(O)(CH2)2C6H4OH
96.5
ethyle3-(4-hydroxyphenyl)propanoate
Amber Glass Bottle
25g
100.0
C11H14O3
MFCD00020209
97%
23795-02-0

Chemical Identifiers

23795-02-0
100.0
C11H14O3
194.23
ethyle3-(4-hydroxyphenyl)propanoate
96.5
97%
CH3CH2OC(O)(CH2)2C6H4OH
MFCD00020209
97%