missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-nitrobenzoylacetate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 430040010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 2-nitrobenzoylacetate | |
35.0°C to 36.0°C | |
Authentic | |
Glass bottle | |
MFCD03424821 | |
ethyl 3-2-nitrophenyl-3-oxopropanoate, ethyl 2-nitrobenzoylacetate, acmc-1awhb, ethyl 3-2-nitrophenyl-3-oxo-propanoate, ethyl o-nitrobenzoylacetate, ethyl 2-o-nitrobenzoyl acetate, ethyl 2-2-nitrobenzoyl acetate, ethyl 2-o-nitrobenzoyl-acetate, ethyl 2-nitrobenzoyl acetate | |
CCOC(=O)CC(=O)C1=CC=CC=C1[N+](=O)[O-] | |
237.21 | |
237.21 | |
Liquid |
52119-39-8 | |
Orange to Brown | |
96% min. (HPLC) | |
C11H11NO5 | |
1 g | |
OWZNCVIBJQPNEF-UHFFFAOYSA-N | |
ethyl 3-(2-nitrophenyl)-3-oxopropanoate | |
10561837 | |
97% |
Chemical Identifiers
52119-39-8 | |
237.21 | |
OWZNCVIBJQPNEF-UHFFFAOYSA-N | |
10561837 | |
CCOC(=O)CC(=O)C1=CC=CC=C1[N+](=O)[O-] |
C11H11NO5 | |
MFCD03424821 | |
ethyl 3-2-nitrophenyl-3-oxopropanoate, ethyl 2-nitrobenzoylacetate, acmc-1awhb, ethyl 3-2-nitrophenyl-3-oxo-propanoate, ethyl o-nitrobenzoylacetate, ethyl 2-o-nitrobenzoyl acetate, ethyl 2-2-nitrobenzoyl acetate, ethyl 2-o-nitrobenzoyl-acetate, ethyl 2-nitrobenzoyl acetate | |
ethyl 3-(2-nitrophenyl)-3-oxopropanoate |
RUO – Research Use Only