missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-methylnicotinate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436190010
Specifications
Ethyl 2-methylnicotinate | |
103°C | |
Glass Bottle | |
126°C to 127°C (24.0mmHg) | |
1g | |
96% min. (GC) | |
MFCD00006336 | |
TZORNSWZUZDUCU-UHFFFAOYSA-N | |
ethyl 2-methylpyridine-3-carboxylate | |
74401 | |
Liquid |
1.0700g/mL | |
Authentic | |
1.07 | |
Colorless to Orange | |
1721-26-2 | |
C9H11NO2 | |
2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 2-methylnicotinic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, ethyl 2-methylnicotinate, ethyl 2-methylpyridine-3-carboxylate~2-methylnicotinic acid ethyl ester, pubchem12727 | |
CCOC(=O)C1=C(N=CC=C1)C | |
165.19 | |
165.19 | |
97% |
Chemical Identifiers
1721-26-2 | |
165.19 | |
TZORNSWZUZDUCU-UHFFFAOYSA-N | |
74401 | |
CCOC(=O)C1=C(N=CC=C1)C |
C9H11NO2 | |
MFCD00006336 | |
2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 2-methylnicotinic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, ethyl 2-methylnicotinate, ethyl 2-methylpyridine-3-carboxylate~2-methylnicotinic acid ethyl ester, pubchem12727 | |
ethyl 2-methylpyridine-3-carboxylate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 217-013-4