missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-methylnicotinate, 97%, ACROS Organics™

Chemical Identifiers

CAS 1721-26-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00006336
InChI Key TZORNSWZUZDUCU-UHFFFAOYSA-N
Synonym 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 2-methylnicotinic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, ethyl 2-methylnicotinate, ethyl 2-methylpyridine-3-carboxylate~2-methylnicotinic acid ethyl ester, pubchem12727
PubChem CID 74401
IUPAC Name ethyl 2-methylpyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC436190010
SDS View Documents
Thermo Fisher Scientific
AC436190010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436190050
SDS View Documents
Thermo Fisher Scientific
AC436190050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1721-26-2
165.19
TZORNSWZUZDUCU-UHFFFAOYSA-N
74401
CCOC(=O)C1=C(N=CC=C1)C
C9H11NO2
MFCD00006336
2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 2-methylnicotinic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, ethyl 2-methylnicotinate, ethyl 2-methylpyridine-3-carboxylate~2-methylnicotinic acid ethyl ester, pubchem12727
ethyl 2-methylpyridine-3-carboxylate

Specifications

1.0700g/mL
Authentic
1.07
Colorless to Orange
1721-26-2
C9H11NO2
2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 2-methylnicotinic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, ethyl 2-methylnicotinate, ethyl 2-methylpyridine-3-carboxylate~2-methylnicotinic acid ethyl ester, pubchem12727
CCOC(=O)C1=C(N=CC=C1)C
165.19
165.19
97%
103°C
Glass Bottle
126°C to 127°C (24.0mmHg)
1g
96% min. (GC)
MFCD00006336
TZORNSWZUZDUCU-UHFFFAOYSA-N
ethyl 2-methylpyridine-3-carboxylate
74401
Liquid
Ethyl 2-methylnicotinate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 217-013-4

SDS