Please login to your online account to display your discounted pricing

ethyle2-methyl-1,3-thiazole-4-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material ethyle2-methyl-1, 3-thiazole-4-carboxylate
CAS 6436-59-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H9NO2S
Formula Weight 171.22
MDL Number MFCD00156148
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB02288DA

  SDS
fisher scientific company
SPB02288DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.22
Quantity 1g
Assay 97%
CAS Min % 97.0
Molecular Formula C7H9NO2S
MDL Number MFCD00156148
Chemical Name or Material ethyle2-methyl-1, 3-thiazole-4-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 6436-59-5