Please login to your online account to display your discounted pricing

ethyle2-methyl-1,3-thiazole-4-carboxylate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB02288DA

 View more versions of this product

Catalog No. SPB02288DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 171.22
Quantity 1g
Assay 97%
CAS Min % 97.0
Molecular Formula C7H9NO2S
MDL Number MFCD00156148
Chemical Name or Material ethyle2-methyl-1, 3-thiazole-4-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 6436-59-5

Specifications

Chemical Name or Material ethyle2-methyl-1, 3-thiazole-4-carboxylate
CAS 6436-59-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H9NO2S
Formula Weight 171.22
MDL Number MFCD00156148