missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
ethyle2-mercapto-1H-imidazole-4-carboxylate, 97%, Thermo Scientific™
Chemical Identifiers
CAS
64038-64-8
Molecular Formula
C6H8N2O2S
Molecular Weight (g/mol)
172.202
MDL Number
MFCD00068042
InChI Key
PONOGPICUOALRU-UHFFFAOYSA-N
Synonym
ethyl 2-mercapto-1h-imidazole-4-carboxylate, 4-ethoxycarbonylimidazole-2-thiol, usaf el-63, imidazole-2-thiol, 5-ethoxycarbonyl, 2-mercapto-5-carbethoxyimidazole, imidazole, 2-mercapto-5-carbethoxy, ethyl 2-sulfanyl-1h-imidazole-4-carboxylate, 2-mercapto-1h-imidazole-4-carboxylic acid ethyl ester, ethyl 2-mercaptoimidazole-4-carboxylate, maybridge1_005066
PubChem CID
2736338
IUPAC Name
ethyl 2-sulfanylidene-1,3-dihydroimidazole-4-carboxylate
SMILES
CCOC(=O)C1=CNC(=S)N1
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
CAS
64038-64-8
Molecular Weight (g/mol)
172.202
InChI Key
PONOGPICUOALRU-UHFFFAOYSA-N
PubChem CID
2736338
SMILES
CCOC(=O)C1=CNC(=S)N1
Molecular Formula
C6H8N2O2S
MDL Number
MFCD00068042
Synonym
ethyl 2-mercapto-1h-imidazole-4-carboxylate, 4-ethoxycarbonylimidazole-2-thiol, usaf el-63, imidazole-2-thiol, 5-ethoxycarbonyl, 2-mercapto-5-carbethoxyimidazole, imidazole, 2-mercapto-5-carbethoxy, ethyl 2-sulfanyl-1h-imidazole-4-carboxylate, 2-mercapto-1h-imidazole-4-carboxylic acid ethyl ester, ethyl 2-mercaptoimidazole-4-carboxylate, maybridge1_005066
IUPAC Name
ethyl 2-sulfanylidene-1,3-dihydroimidazole-4-carboxylate
Specifications
CAS
64038-64-8
Assay Percent Range
97%
Molecular Formula
C6H8N2O2S
Quantity
1g
InChI Key
PONOGPICUOALRU-UHFFFAOYSA-N
IUPAC Name
ethyl 2-sulfanylidene-1,3-dihydroimidazole-4-carboxylate
PubChem CID
2736338
Percent Purity
97%
CAS Min %
97.0
Packaging
Amber Glass Bottle
MDL Number
MFCD00068042
Synonym
ethyl 2-mercapto-1h-imidazole-4-carboxylate, 4-ethoxycarbonylimidazole-2-thiol, usaf el-63, imidazole-2-thiol, 5-ethoxycarbonyl, 2-mercapto-5-carbethoxyimidazole, imidazole, 2-mercapto-5-carbethoxy, ethyl 2-sulfanyl-1h-imidazole-4-carboxylate, 2-mercapto-1h-imidazole-4-carboxylic acid ethyl ester, ethyl 2-mercaptoimidazole-4-carboxylate, maybridge1_005066
SMILES
CCOC(=O)C1=CNC(=S)N1
Molecular Weight (g/mol)
172.202
Formula Weight
172.21
Chemical Name or Material
Ethyl 2-mercapto-1H-imidazole-4-carboxylate
Safety and Handling
Safety and Handling
GHS H Statement: Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'