Please login to your online account to display your discounted pricing

ethyle2-chloro-3-cyano-6-methylisonicotinate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material ethyle2-chloro-3-cyano-6-methylisonicotinate
CAS 40108-12-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9ClN2O2
Formula Weight 224.65
MDL Number MFCD00023416
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF00966CB

  SDS
fisher scientific company
RF00966CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.65
Quantity 250mg
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
CAS 40108-12-1
Molecular Formula C10H9ClN2O2
Assay Percent Range 97%
MDL Number MFCD00023416
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material ethyle2-chloro-3-cyano-6-methylisonicotinate