missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-butynoate, 99%, ACROS Organics™
$39.77 - $194.19
Chemical Identifiers
CAS | 4341-76-8 |
---|---|
Molecular Formula | C6H8O2 |
Molecular Weight (g/mol) | 112.13 |
MDL Number | MFCD00015182 |
InChI Key | FCJJZKCJURDYNF-UHFFFAOYSA-N |
Synonym | 2-butynoic acid ethyl ester, 2-butynoic acid, ethyl ester, acmc-1aeo2, but-2-ynoic acid ethyl ester, ethyl 1-propynecarboxylate, ethyl 2-butynoate, ethyl 3-methylpropiolate, ethyl tetrolate, ethyl-2-butynoate, tetrolic acid ethyl ester |
PubChem CID | 78043 |
IUPAC Name | ethyl but-2-ynoate |
SMILES | CCOC(=O)C#CC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC430030010
|
Thermo Fisher Scientific
AC430030010 |
1g | Glass bottle |
Each for $39.77
|
|
AC430030100
|
Thermo Fisher Scientific
AC430030100 |
10g | Glass bottle |
Each for $194.19
|
|
Chemical Identifiers
4341-76-8 | |
112.13 | |
FCJJZKCJURDYNF-UHFFFAOYSA-N | |
78043 | |
CCOC(=O)C#CC |
C6H8O2 | |
MFCD00015182 | |
2-butynoic acid ethyl ester, 2-butynoic acid, ethyl ester, acmc-1aeo2, but-2-ynoic acid ethyl ester, ethyl 1-propynecarboxylate, ethyl 2-butynoate, ethyl 3-methylpropiolate, ethyl tetrolate, ethyl-2-butynoate, tetrolic acid ethyl ester | |
ethyl but-2-ynoate |
Specifications
58°C | |
Glass bottle | |
Brown to Gray | |
4341-76-8 | |
C6H8O2 | |
2-butynoic acid ethyl ester, 2-butynoic acid, ethyl ester, acmc-1aeo2, but-2-ynoic acid ethyl ester, ethyl 1-propynecarboxylate, ethyl 2-butynoate, ethyl 3-methylpropiolate, ethyl tetrolate, ethyl-2-butynoate, tetrolic acid ethyl ester | |
CCOC(=O)C#CC | |
112.13 | |
112.13 | |
99% |
Authentic | |
160.0°C to 164.0°C | |
1g | |
98.5% min. (GC) | |
MFCD00015182 | |
FCJJZKCJURDYNF-UHFFFAOYSA-N | |
ethyl but-2-ynoate | |
78043 | |
Powder | |
Ethyl 2-butynoate |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Warning
EINECSNumber : 224-395-6