Please login to your online account to display your discounted pricing

ethyle2-bromobenzoate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material ethyle2-bromobenzoate
CAS 6091-64-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9BrO2
Linear Formula BrC6H4CO2C2H5
Formula Weight 229.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDP00160EA

  SDS
fisher scientific company
RDP00160EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.07
Quantity 10g
Assay 97%
Density 1.44g/mL
CAS Min % 97.0
Chemical Name or Material ethyle2-bromobenzoate
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C9H9BrO2
Linear Formula BrC6H4CO2C2H5
CAS 6091-64-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.