missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle2-amino-5-methyl-4-phenylthiophene-3-carboxylate, 97%, Maybridge

Manufacturer:  Fisher Scientific Company AW00937EB

 View more versions of this product

Catalog No. AW00937EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

ethyle2-amino-5-methyl-4-phenylthiophene-3-carboxylate
4815-37-6
100.0
C14H15NO2S
2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl ester, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, maybridge1_000503, upcmld-dp080, upcmld-dp080:001
CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N
261.339
261.34
25g
96.5
97%
MFCD01763475
SXONDAWSQJWZEO-UHFFFAOYSA-N
ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
693902
97%

Chemical Identifiers

4815-37-6
261.339
SXONDAWSQJWZEO-UHFFFAOYSA-N
693902
CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N
C14H15NO2S
MFCD01763475
2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl ester, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, maybridge1_000503, upcmld-dp080, upcmld-dp080:001
ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents