Please login to your online account to display your discounted pricing

ethyle2-amino-4-(4-bromophenyl)-3-thiophenecarboxylate, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  CC00243EA

 View more versions of this product

Catalog No. CC00243EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 326.21
Quantity 10g
Assay 97%
CAS Max % 100.0
CAS 306934-99-6
Chemical Name or Material Ethyl 2-amino-4-(4-bromophenyl)-3-thiophenecarboxylate
Assay Percent Range 97%
Molecular Formula C13H12BrNO2S
MDL Number MFCD00435056
CAS Min % 96.5

Specifications

Chemical Name or Material Ethyl 2-amino-4-(4-bromophenyl)-3-thiophenecarboxylate
CAS 306934-99-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H12BrNO2S
Formula Weight 326.21
MDL Number MFCD00435056