missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle2-(5-bromo-2-thienyl)-2-oxoacetate, 97%, Maybridge

Manufacturer:  Fisher Scientific Company BTB09133EE

 View more versions of this product

Catalog No. BTB09133EE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

263.11
97%
Ethyl 2-(5-bromo-2-thienyl)-2-oxoacetate
96.5
C8H7BrO3S
50g
100.0
MFCD00085056
22098-10-8
97%

Chemical Identifiers

22098-10-8
100.0
C8H7BrO3S
MFCD00085056
97%
96.5
97%
263.11
Ethyl 2-(5-bromo-2-thienyl)-2-oxoacetate

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.