Please login to your online account to display your discounted pricing

ethyle2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate
CAS 61713-23-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H16O4S2
Linear Formula CH3CH2O2CCH2SCH2SCH2CO2CH2CH3
Formula Weight 252.35
MDL Number MFCD00026901
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S12083DA

 
fisher scientific company
S12083DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.35
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate
Assay Percent Range 97%
CAS 61713-23-3
Packaging Amber glass bottle
Molecular Formula C9H16O4S2
Linear Formula CH3CH2O2CCH2SCH2SCH2CO2CH2CH3
MDL Number MFCD00026901