Please login to your online account to display your discounted pricing

ethyle2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  S12083DA

 View more versions of this product

Catalog No. S12083DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 252.35
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate
CAS 61713-23-3
Assay Percent Range 97%
MDL Number MFCD00026901
Linear Formula CH3CH2O2CCH2SCH2SCH2CO2CH2CH3
Packaging Amber glass bottle
Molecular Formula C9H16O4S2

Specifications

Chemical Name or Material Ethyl 2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate
CAS 61713-23-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H16O4S2
Linear Formula CH3CH2O2CCH2SCH2SCH2CO2CH2CH3
Formula Weight 252.35
MDL Number MFCD00026901