Please login to your online account to display your discounted pricing

ethyle1-methylcyclopropane-1-carboxylate, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  SB01040EA

 View more versions of this product

Catalog No. SB01040EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 128.17
Quantity 10g
Assay 97%
Density 0.9180g/mL
CAS Min % 97.0
Chemical Name or Material Ethyl 1-methylcyclopropane-1-carboxylate
Assay Percent Range 97%
Linear Formula CH3C3H4C(O)OCH2CH3
MDL Number MFCD00001284
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7H12O2
CAS 71441-76-4
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.

Specifications

Chemical Name or Material Ethyl 1-methylcyclopropane-1-carboxylate
CAS 71441-76-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H12O2
Linear Formula CH3C3H4C(O)OCH2CH3
Formula Weight 128.17
MDL Number MFCD00001284
Accessories

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents