Please login to your online account to display your discounted pricing

ethyle1-benzylpiperidine-4-carboxylate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM09611DA

 View more versions of this product

Catalog No. KM09611DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 247.34
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
MDL Number MFCD00040748
Chemical Name or Material Ethyl 1-benzylpiperidine-4-carboxylate
Assay Percent Range 97%
Molecular Formula C15 H21 N O2
CAS 24228-40-8

Specifications

Chemical Name or Material Ethyl 1-benzylpiperidine-4-carboxylate
CAS 24228-40-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15 H21 N O2
Formula Weight 247.34
MDL Number MFCD00040748