missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle1-benzylpiperidine-4-carboxylate, 97%, Maybridge

$52.08 - $52.08

Chemical Identifiers

CAS 24228-40-8
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C15 H21 N O2
Formula Weight 247.34
MDL Number MFCD00040748
Chemical Name or Material Ethyl 1-benzylpiperidine-4-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
KM09611DA SDS Fisher Scientific Company
KM09611DA
Amber Glass Bottle 1g Each for $52.08
Specifications Description & Specifications

Chemical Identifiers

24228-40-8
97%
247.34
Ethyl 1-benzylpiperidine-4-carboxylate
97.0
C15 H21 N O2
MFCD00040748
97%

Specifications

247.34
97.0
MFCD00040748
24228-40-8
Ethyl 1-benzylpiperidine-4-carboxylate
1g
Amber Glass Bottle
97%
C15 H21 N O2
97%