Please login to your online account to display your discounted pricing

ethyle1,4-dibenzylpiperazine-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 1, 4-dibenzylpiperazine-2-carboxylate
CAS 72351-59-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C21H26N2O2
Formula Weight 338.45
MDL Number MFCD00126366
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM07907DA

 
fisher scientific company
KM07907DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 338.45
Quantity 1g
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Molecular Formula C21H26N2O2
CAS 72351-59-8
Chemical Name or Material Ethyl 1, 4-dibenzylpiperazine-2-carboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00126366
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.