Please login to your online account to display your discounted pricing

ethyle1-(4-cyanophenyl)-4-piperidinecarboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 1-(4-cyanophenyl)-4-piperidinecarboxylate
CAS 352018-90-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H18N2O2
Formula Weight 258.32
MDL Number MFCD03086198
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00017EA

 
fisher scientific company
MO00017EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.32
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 1-(4-cyanophenyl)-4-piperidinecarboxylate
Health Hazard 1 GHS Signal Word: Warning
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C15H18N2O2
MDL Number MFCD03086198
CAS 352018-90-7
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.