Please login to your online account to display your discounted pricing

ethyle1-(4-cyanophenyl)-4-piperidinecarboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00017EA

 View more versions of this product

Catalog No. MO00017EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 258.32
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Ethyl 1-(4-cyanophenyl)-4-piperidinecarboxylate
Health Hazard 1 GHS Signal Word: Warning
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C15H18N2O2
MDL Number MFCD03086198
CAS 352018-90-7
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ethyl 1-(4-cyanophenyl)-4-piperidinecarboxylate
CAS 352018-90-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H18N2O2
Formula Weight 258.32
MDL Number MFCD03086198
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

SDS & Safety Documents