Please login to your online account to display your discounted pricing

ethyle(S)-3-hydroxybutyrate, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC335260050

 View more versions of this product

Catalog No. AC335260050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 132.16
Quantity 5g
Color Clear colorless
Assay 99%
Specific Rotation Condition + 43.00 (20.00°C c=1,chloroform)
Density 1.0100g/mL
Packaging Glass bottle
Specific Rotation +43° to +47° (20°C, 589nm) (c=1, CHCl3)
CAS 56816-01-4
Assay Percent Range 99%
Chemical Name or Material Ethyl (S)-3-hydroxybutyrate
Molecular Formula C6H12O3
MDL Number MFCD00066206
Refractive Index 1.4195 to 1.4215 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula CH3CH(OH)CH2COOCH2CH3

Specifications

Chemical Name or Material Ethyl (S)-3-hydroxybutyrate
Assay 99%
Assay Percent Range 99%
Molecular Formula C6H12O3
Linear Formula CH3CH(OH)CH2COOCH2CH3
Formula Weight 132.16
MDL Number MFCD00066206