missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle(S)-3-hydroxybutyrate, 99%, ACROS Organics™

Chemical Identifiers

CAS 56816-01-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00066206
InChI Key OMSUIQOIVADKIM-YFKPBYRVSA-N
Synonym s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
PubChem CID 6950306
IUPAC Name ethyl (3S)-3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC335260010 SDS View Documents Acros Organics
335260010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

56816-01-4
132.159
OMSUIQOIVADKIM-YFKPBYRVSA-N
6950306
CCOC(=O)CC(C)O
C6H12O3
MFCD00066206
s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
ethyl (3S)-3-hydroxybutanoate

Specifications

1.0100g/mL
Glass bottle
+ 43.00 (20.00°C c=1,chloroform)
Colorless
56816-01-4
C6H12O3
MFCD00066206
OMSUIQOIVADKIM-YFKPBYRVSA-N
ethyl (3S)-3-hydroxybutanoate
6950306
Liquid
Ethyl (S)-3-hydroxybutyrate
Authentic
1.4195 to 1.4215 (20°C, 589nm)
+43° to +47° (20°C, 589nm) (c=1, CHCl3)
1g
99%
CH3CH(OH)CH2COOCH2CH3
s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
CCOC(=O)CC(C)O
132.159
132.16
99%