Please login to your online account to display your discounted pricing

ethyle(S)-3-hydroxybutyrate, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  335260010

 View more versions of this product

Catalog No. AC335260010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 132.16
Quantity 1g
Color Clear colorless
Density 1.0100g/mL
Assay 99%
Specific Rotation Condition + 43.00 (20.00°C c=1,chloroform)
Packaging Glass bottle
Chemical Name or Material Ethyl (S)-3-hydroxybutyrate
Assay Percent Range 99%
Molecular Formula C6H12O3
Refractive Index 1.4195 to 1.4215 (20°C, 589nm)
MDL Number MFCD00066206
CAS 56816-01-4
Infrared Spectrum Authentic
Linear Formula CH3CH(OH)CH2COOCH2CH3
Specific Rotation +43° to +47° (20°C, 589nm) (c=1, CHCl3)

Specifications

Chemical Name or Material Ethyl (S)-3-hydroxybutyrate
Assay 99%
Assay Percent Range 99%
Molecular Formula C6H12O3
Linear Formula CH3CH(OH)CH2COOCH2CH3
Formula Weight 132.16
MDL Number MFCD00066206