Please login to your online account to display your discounted pricing

Ethyldiphenylphosphine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyldiphenylphosphine
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14H15P
Linear Formula (C6H5)2PCH2CH3
Formula Weight 214.25
MDL Number MFCD00015170
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383010050

  SDS
thermo fisher scientific
AC383010050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC383010010

  SDS
thermo fisher scientific
AC383010010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 214.25
Quantity 5g
Color Clear colorless
Density 1.04g/mL
Chemical Name or Material Ethyldiphenylphosphine
Refractive Index 1.6130 to 1.6150 (20°C, 589nm)
Molecular Formula C14H15P
CAS 607-01-2
Packaging Glass bottle
Assay Percent Range 99%
MDL Number MFCD00015170
Linear Formula (C6H5)2PCH2CH3
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.