Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyldimethylchlorosilane, 97%

Chemical Identifiers

Chemical Name or Material Ethyldimethylchlorosilane
CAS 6917-76-6
Molecular Formula CH3CH2(CH3)2SiCl
Formula Weight 122.67
MDL Number MFCD00009918
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1322214

  SDS
alfa aesar™
A1322214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.67
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula CH3CH2(CH3)2SiCl
Assay 97%
MDL Number MFCD00009918
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2924
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Chemical Name or Material Ethyldimethylchlorosilane
Health Hazard 1 Danger
Boiling Point 90° to 91°C
Quantity 25g
CAS 6917-76-6
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.