missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylcyclohexane, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 118311000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethylcyclohexane | |
1678-91-7 | |
130°C to 132°C | |
Authentic | |
Glass bottle | |
1.4311 to 1.4331 | |
MFCD00001523 | |
05, 35 | |
cyclohexane, ethyl, ethyl cyclohexane, cyclohexylethane, ethyl-cyclohexane, acmc-1c1qu, ethylcyclohexane, dsstox_cid_30334, dsstox_gsid_51779, wln: l6tj a2, qspl 100 | |
IIEWJVIFRVWJOD-UHFFFAOYSA-N | |
ethylcyclohexane | |
15504 | |
99+% |
99+% | |
0.7800g/mL | |
18°C | |
99+% | |
C8H16 | |
C6H11C2H5 | |
100mL | |
0.78 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,carbon tetrachloride,lignoin | |
CCC1CCCCC1 | |
112.21 | |
112.21 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 216-835-0
TSCA : TSCA
RUO â Research Use Only