missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylcyclohexane, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 118310250
Specifications
Ethylcyclohexane | |
18°C | |
Glass bottle | |
0.78 | |
Colorless | |
25mL | |
1678-91-7 | |
C8H16 | |
MFCD00001523 | |
acmc-1c1qu, cyclohexane, ethyl, cyclohexylethane, dsstox_cid_30334, dsstox_gsid_51779, ethyl cyclohexane, ethyl-cyclohexane, ethylcyclohexane, qspl 100, wln: l6tj a2 | |
IIEWJVIFRVWJOD-UHFFFAOYSA-N | |
ethylcyclohexane | |
15504 | |
Liquid |
0.7800g/mL | |
Authentic | |
1.4311 to 1.4331 | |
130°C to 132°C | |
-111°C | |
99+% | |
99+% | |
C6H11C2H5 | |
05, 35 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,carbon tetrachloride,lignoin | |
CCC1CCCCC1 | |
112.21 | |
112.21 | |
99+% |
Chemical Identifiers
1678-91-7 | |
112.21 | |
IIEWJVIFRVWJOD-UHFFFAOYSA-N | |
15504 | |
CCC1CCCCC1 |
C8H16 | |
MFCD00001523 | |
acmc-1c1qu, cyclohexane, ethyl, cyclohexylethane, dsstox_cid_30334, dsstox_gsid_51779, ethyl cyclohexane, ethyl-cyclohexane, ethylcyclohexane, qspl 100, wln: l6tj a2 | |
ethylcyclohexane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 216-835-0
TSCA : TSCA