missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 118080251

 View more versions of this product

Catalog No. AC118080251



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethylbenzene
100-41-4
0.8600g/mL
22°C
99.7% min. (GC)
C8H10
C6H5C2H5
25 L
0.86
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
ethylbenzene
7500
CHEBI:16101
99.8%
Liquid
99.8%
-95.0°C
136.0°C
Authentic
Metal Drum
1.4940 to 1.4970
MFCD00011647
05, II, 274
15, 3820
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, benzene, carbon tetrachloride,, ether and sulfur dioxide, slightly soluble in chloroform, insoluble in ammonia
CCC1=CC=CC=C1
106.17
0.63 mPa.s (20°C)
106.17
Pure

Chemical Identifiers

100-41-4
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
7500
ethylbenzene
MFCD00011647
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
CHEBI:16101
CCC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasti

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Danger

EINECSNumber : 202-849-4

RTECSNumber : DA0700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only