Please login to your online account to display your discounted pricing

Ethylaluminium dichloride, 1.8M solution in toluene, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC428048000

 View more versions of this product

Catalog No. AC428048000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 126.95
Physical Form Liquid
Quantity 800mL
Color Clear colorless to yellow
Merck Index 15, 3816
Packaging AcroSeal™ Glass Bottle
Density 0.934g/mL
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging the unborn child.
In contact with water rele
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Name Note 1.8M solution in toluene
Molecular Formula C2H5AlCl2
Chemical Name or Material Ethylaluminium dichloride
CAS 563-43-9
MDL Number MFCD00000457
Linear Formula C2H5AlCl2
CAS 108-88-3

Specifications

Chemical Name or Material Ethylaluminium dichloride
Name Note 1.8M solution in toluene
CAS 563-43-9
CAS 108-88-3
Molecular Formula C2H5AlCl2
Linear Formula C2H5AlCl2
Formula Weight 126.95
MDL Number MFCD00000457
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging the unborn child.
In contact with water rele

SDS & Safety Documents