Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl xanthene-9-carboxylate, 98%

$158.22 - $452.03

Chemical Identifiers

Chemical Name or Material Ethyl xanthene-9-carboxylate
CAS 7401-03-8
Molecular Formula C16H13O3
Formula Weight 254.29
MDL Number MFCD00100669
Synonym Xanthene-9-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1709506

  SDS
alfa aesar™
A1709506
5g Each for $158.22

AAA1709514

  SDS
alfa aesar™
A1709514
25g Each for $452.03
Description & Specifications

Specifications

Formula Weight 254.29
Melting Point 66° to 68°C
Chemical Name or Material Ethyl xanthene-9-carboxylate
Molecular Formula C16H13O3
Assay 98%
CAS 7401-03-8
Synonym Xanthene-9-carboxylic acid ethyl ester
Quantity 5g
MDL Number MFCD00100669