missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl vinyl ketone, 98%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 119090050

 View more versions of this product

Catalog No. AC119090050



Description

Description

Specifications

Specifications

Ethyl vinyl ketone
-6°C
Glass bottle
0.849
103°C to 104°C
5g
1629-58-9,128-37-0
C5H8O
MFCD00009316
01,388
JLIDVCMBCGBIEY-UHFFFAOYSA-N
pent-1-en-3-one
15394
Liquid
0.8490g/mL
Authentic
1.4180 to 1.422
0.1% (BHT)
Amber to Colorless
Stabilized
97.5% min. (GC)
C2H5COCH=CH2
01, 731
1-penten-3-one, 1-pentene-3-one, ethyl vinyl ketone, ethylvinyl ketone, ethylvinylketone, fema no. 3382, ketone, ethyl vinyl, pentenone, unii-r0053y1az7, vinyl ethyl ketone
CCC(=O)C=C
84.12
84.12
98%

Chemical Identifiers

1629-58-9
84.12
JLIDVCMBCGBIEY-UHFFFAOYSA-N
15394
CCC(=O)C=C
C5H8O
MFCD00009316
1-penten-3-one, 1-pentene-3-one, ethyl vinyl ketone, ethylvinyl ketone, ethylvinylketone, fema no. 3382, ketone, ethyl vinyl, pentenone, unii-r0053y1az7, vinyl ethyl ketone
pent-1-en-3-one
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.

GHS Signal Word: Danger

EINECSNumber : 216-624-3

SDS Documents