Please login to your online account to display your discounted pricing

Ethyl Vinyl Ketone, Stabilized, 98%, ACROS Organics™

$55.01 - $201.91

Chemical Identifiers

Chemical Name or Material Ethyl vinyl ketone
Name Note Stabilized
CAS 1629-58-9
CAS 128-37-0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H8O
Linear Formula C2H5COCH=CH2
Formula Weight 84.12
MDL Number MFCD00009316
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119090050

  SDS
thermo fisher scientific
119090050
Glass bottle 5g Each for $55.01

AC119090250

  SDS
thermo fisher scientific
AC119090250
Glass bottle 25g Each for $201.91
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 84.12
Quantity 5g
Color Clear colorless to amber
Beilstein 01, 731
Fieser 01,388
Density 0.8490g/mL
Linear Formula C2H5COCH=CH2
CAS 1629-58-9
Synonym EVK
Packaging Glass bottle
Chemical Name or Material Ethyl vinyl ketone
Refractive Index 1.4180 to 1.4220 (20°C, 589nm)
Stabilizer 0.1% (BHT)
Name Note Stabilized
Assay Percent Range 98%
Molecular Formula C5H8O
MDL Number MFCD00009316
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Highly flammable liquid and vapor.
CAS 128-37-0
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
Infrared Spectrum Authentic
Safety and Handling

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

GHS P Statement Highly flammable liquid and vapor.