Please login to your online account to display your discounted pricing

Ethyl Vinyl Ketone, Stabilized, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC119090250

 View more versions of this product

Catalog No. AC119090250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 84.12
Quantity 25g
Color Clear colorless to amber
Density 0.8490g/mL
Beilstein 01, 731
Fieser 01,388
MDL Number MFCD00009316
CAS 1629-58-9
Synonym EVK
CAS 128-37-0
Chemical Name or Material Ethyl vinyl ketone
Refractive Index 1.4180 to 1.4220 (20°C, 589nm)
Stabilizer 0.1% (BHT)
Name Note Stabilized
Assay Percent Range 98%
Molecular Formula C5H8O
Linear Formula C2H5COCH=CH2
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Ethyl vinyl ketone
Name Note Stabilized
CAS 1629-58-9
CAS 128-37-0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H8O
Linear Formula C2H5COCH=CH2
Formula Weight 84.12
MDL Number MFCD00009316
Synonym EVK
Accessories

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

GHS P Statement Highly flammable liquid and vapor.

SDS & Safety Documents