missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl vinyl ether, 99%, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 119080010
Specifications
Ethyl vinyl ether | |
−45°C | |
Glass bottle | |
0.75 | |
0.28 mPa.s (5°C) | |
Colorless | |
1L | |
109-92-2,1310-58-3 | |
C4H8O | |
MFCD00009248 | |
01,386; 02,198; 04,234; 10,184; 11,235 | |
Solubility in water: 7.8g/L (25°C). Other solubilities: soluble in most organic solvents | |
CCOC=C | |
72.11 | |
72.11 | |
99% |
0.7500g/mL | |
Authentic | |
1.3760 to 1.3780 | |
0.1% KOH | |
36.0°C | |
-116.0°C | |
Stabilized | |
98.5% min. (GC) | |
C2H5OCH=CH2 | |
01,433 | |
1-ethoxyethene, 1-ethoxyethylene, ethene, ethoxy, ether, ethyl vinyl, ether, vinyl ethyl, ethoxyethene, ethoxyethylene, ethyl vinyl ether, vinamar, vinyl ethyl ether | |
FJKIXWOMBXYWOQ-UHFFFAOYSA-N | |
ethenoxyethane | |
8023 | |
Liquid |
Chemical Identifiers
109-92-2 | |
72.11 | |
FJKIXWOMBXYWOQ-UHFFFAOYSA-N | |
8023 | |
CCOC=C |
C4H8O | |
MFCD00009248 | |
1-ethoxyethene, 1-ethoxyethylene, ethene, ethoxy, ether, ethyl vinyl, ether, vinyl ethyl, ethoxyethene, ethoxyethylene, ethyl vinyl ether, vinamar, vinyl ethyl ether | |
ethenoxyethane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Danger
EINECSNumber : 203-718-4