missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl vinyl ether, 99%, stab.

Manufacturer:  Alfa Aesar™ A156910F

 View more versions of this product

Catalog No. AAA156910F



Description

Description

Applications
Ethyl vinyl ether is used in fragrances, lubricating oil additives and spices preparation. It acts as an intermediate for sulfadiazine. It finds application in pharmaceuticals as anesthetics and analgesics. Further, it is used as a solvent in organic synthesis.

Solubility
Miscible with common organic solvents. Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, oxygen and acids. Store in a cool place.
Specifications

Specifications

Ethyl vinyl ether
Ether-like
-115°C
stabilized
C4H8O
MFCD00009248
605351
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
ethenoxyethane
8023
99%
−45°C (−49°F)
33°C to 34°C
2500mL
109-92-2
CH2=CHOCH2CH3
UN1302
ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl
CCOC=C
72.107
72.11

Chemical Identifiers

109-92-2
72.107
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
8023
CCOC=C
C4H8O
MFCD00009248
ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl
ethenoxyethane
Safety and Handling

Safety and Handling

GHS H Statement
H224-H319-H335-H336
Extremely flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: VINYL ETHYL ETHER, STABILIZED

RTECSNumber : KO0710000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents