missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl valerate, 98%

Manufacturer:  Alfa Aesar™ A14854AE

 View more versions of this product

Catalog No. AAA14854AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl valerate
142°C to 145°C
100mL
C7H14O2
UN3272
14,9904
ICMAFTSLXCXHRK-UHFFFAOYSA-N
ethyl pentanoate
10882
98%
38°C (100°F)
-92°C to -90°C
539-82-2
MFCD00009479
1744680
ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
CCCCC(=O)OCC
130.187
130.19

Chemical Identifiers

539-82-2
130.187
ICMAFTSLXCXHRK-UHFFFAOYSA-N
10882
CCCCC(=O)OCC
C7H14O2
MFCD00009479
ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
ethyl pentanoate
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

SDS Documents