missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl uracil-5-carboxylate, 98%, Thermo Scientific Chemicals
$120.70 - $380.04
Chemical Identifiers
CAS | 28485-17-8 |
---|---|
Molecular Formula | C7H8N2O4 |
Molecular Weight (g/mol) | 184.15 |
MDL Number | MFCD00057337 |
InChI Key | MKNYHTGOVKPZMU-UHFFFAOYSA-N |
Synonym | 5-carbethoxyuracil, ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, isoorotic acid ethyl ester, ethyl 2,4-dihydroxypyrimidine-5-carboxylate, ethyl uracil-5-carboxylate, ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester, ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate, pubchem13254 |
PubChem CID | 101411 |
IUPAC Name | ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate |
SMILES | CCOC(=O)C1=CNC(=O)NC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2650703
|
Thermo Scientific Chemicals
H2650703 |
1 g |
N/A
|
|
|||||
AAH2650706
|
Thermo Scientific Chemicals
H2650706 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
28485-17-8 | |
184.15 | |
MKNYHTGOVKPZMU-UHFFFAOYSA-N | |
101411 | |
CCOC(=O)C1=CNC(=O)NC1=O |
C7H8N2O4 | |
MFCD00057337 | |
5-carbethoxyuracil, ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, isoorotic acid ethyl ester, ethyl 2,4-dihydroxypyrimidine-5-carboxylate, ethyl uracil-5-carboxylate, ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester, ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate, pubchem13254 | |
ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate |
Specifications
28485-17-8 | |
MFCD00057337 | |
5-carbethoxyuracil, ethyl4-hydroxy-2-oxo-1,2-dihydropyrimidine-5-carboxylate, ethyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, isoorotic acid ethyl ester, ethyl 2,4-dihydroxypyrimidine-5-carboxylate, ethyl uracil-5-carboxylate, ethyl 1,2,3,4-tetrahydro-2,4-dioxopyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo-, ethyl ester, ethyl 2,4-dioxo-1,3-dihydropyrimidine-5-carboxylate, pubchem13254 | |
CCOC(=O)C1=CNC(=O)NC1=O | |
184.15 | |
184.15 | |
Ethyl uracil-5-carboxylate |
C7H8N2O4 | |
1 g | |
MKNYHTGOVKPZMU-UHFFFAOYSA-N | |
ethyl 2,4-dioxo-1H-pyrimidine-5-carboxylate | |
101411 | |
98% |
Safety and Handling
EINECSNumber : 249-053-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only