Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl undecanoate, 97%

$42.19 - $128.38

Chemical Identifiers

Chemical Name or Material Ethyl undecanoate
CAS 627-90-7
Molecular Formula C13H26O2
Linear Formula CH3(CH2)9CO2CH2CH3
Formula Weight 214.35
MDL Number MFCD00008958
Beilstein 1767805
Synonym Undecanoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2341614

  SDS
alfa aesar™
B2341614
25g Each for $42.19

AAB2341622

  SDS
alfa aesar™
B2341622
100g Each for $128.38
Description & Specifications

Specifications

Formula Weight 214.35
Chemical Name or Material Ethyl undecanoate
TSCA TSCA
Assay 97%
Molecular Formula C13H26O2
Linear Formula CH3(CH2)9CO2CH2CH3
MDL Number MFCD00008958
CAS 627-90-7
Beilstein 1767805
Synonym Undecanoic acid ethyl ester
Flash Point >110°C (230°F)
Boiling Point 103° to 105°C (4mmHg)
Melting Point −15°C
Quantity 25g