missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyltriphenylphosphonium bromide, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 119040250

 View more versions of this product

Catalog No. AC119040250



Description

Description

for C-C Bond Formation (Olefination)
Specifications

Specifications

Ethyltriphenylphosphonium bromide
Authentic
Glass bottle
206.5°C to 208.5°C
99+%
97.5
97.5% min. (on dry substance) (Argentometry)
C2H5P(C6H5)3Br
ethyl triphenyl phosphonium bromide, ethyl triphenylphosphonium bromide, ethyltriphenylphosphanium bromide, ethyltriphenylphosphonium bromide, ethyltriphenylphosphoniumbromide, guaifenesin powder, phosphonium, ethyltriphenyl-, bromide, pubchem18705, triphenylethylphosphonium bromide, unii-r85v84ul5v
JHYNXXDQQHTCHJ-UHFFFAOYSA-M
ethyl(triphenyl)phosphanium;bromide
73727
Crystalline Powder
>200°C
1% max.
White
25g
1530-32-1
100.0
C20H20BrP
MFCD00011838
Solubility in water: 120g/L (23°C). Other solubilities: soluble in chloroform,fairly soluble in lower alcohols,slightly soluble in arom. hydrocarbons
CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
371.26
371.26
98%

Chemical Identifiers

1530-32-1
371.26
JHYNXXDQQHTCHJ-UHFFFAOYSA-M
73727
CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
C20H20BrP
MFCD00011838
ethyl triphenyl phosphonium bromide, ethyl triphenylphosphonium bromide, ethyltriphenylphosphanium bromide, ethyltriphenylphosphonium bromide, ethyltriphenylphosphoniumbromide, guaifenesin powder, phosphonium, ethyltriphenyl-, bromide, pubchem18705, triphenylethylphosphonium bromide, unii-r85v84ul5v
ethyl(triphenyl)phosphanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/

GHS Signal Word: Danger

EINECSNumber : 216-223-3

TSCA : TSCA

SDS Documents