Learn More
Ethyl (trimethylsilyl)acetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1770730
Description
Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.
Solubility
Decomposes in water.
Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Ethyl (trimethylsilyl)acetate | |
0.876 | |
35°C (95°F) | |
1.415 | |
250 g | |
1755902 | |
ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie | |
QQFBQBDINHJDMN-UHFFFAOYSA-N | |
ethyl 2-trimethylsilylacetate | |
77687 | |
≥98% |
4071-88-9 | |
156°C to 158°C | |
C7H16O2Si | |
MFCD00009172 | |
UN3272 | |
Moisture sensitive | |
Decomposes in water. | |
CCOC(=O)C[Si](C)(C)C | |
160.288 | |
160.29 |
Chemical Identifiers
4071-88-9 | |
160.288 | |
QQFBQBDINHJDMN-UHFFFAOYSA-N | |
77687 | |
CCOC(=O)C[Si](C)(C)C |
C7H16O2Si | |
MFCD00009172 | |
ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie | |
ethyl 2-trimethylsilylacetate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 223-783-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only