missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl trimethylacetopyruvate, 98%

Manufacturer:  Alfa Aesar™ A1516414

 View more versions of this product

Catalog No. AAA1516414



Specifications

Specifications

Ethyl trimethylacetopyruvate
25g
C10H16O4
510374
NIMKIMUBJFWPTD-UHFFFAOYSA-N
ethyl 5,5-dimethyl-2,4-dioxohexanoate
83403
98%
68°C to 70°C (0.5mmHg)
13395-36-3
MFCD00052319
ethyl trimethylacetopyruvate, hexanoic acid, 5,5-dimethyl-2,4-dioxo-, ethyl ester, 5,5-dimethyl-2,4-dioxo-hexanoic acid ethyl ester, 5,5-dimethyl-2,4-dioxohexanoic acid ethyl ester, ethyl pivaloylpyruvate, pivaloylpyruvic acid ethyl ester, 3-pivaloylpyruvic acid ethyl ester, ethyl5,5-dimethyl-2,4-dioxohexanoate, trimethylacetopyruvic acid ethyl ester, ethyl 5,5-dimethyl-2,4-dioxo-hexanoate
CCOC(=O)C(=O)CC(=O)C(C)(C)C
200.234
200.24

Chemical Identifiers

13395-36-3
200.234
NIMKIMUBJFWPTD-UHFFFAOYSA-N
83403
CCOC(=O)C(=O)CC(=O)C(C)(C)C
C10H16O4
MFCD00052319
ethyl trimethylacetopyruvate, hexanoic acid, 5,5-dimethyl-2,4-dioxo-, ethyl ester, 5,5-dimethyl-2,4-dioxo-hexanoic acid ethyl ester, 5,5-dimethyl-2,4-dioxohexanoic acid ethyl ester, ethyl pivaloylpyruvate, pivaloylpyruvic acid ethyl ester, 3-pivaloylpyruvic acid ethyl ester, ethyl5,5-dimethyl-2,4-dioxohexanoate, trimethylacetopyruvic acid ethyl ester, ethyl 5,5-dimethyl-2,4-dioxo-hexanoate
ethyl 5,5-dimethyl-2,4-dioxohexanoate
Documents