Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl trimethylacetate, 99%

Chemical Identifiers

Chemical Name or Material Ethyl trimethylacetate
CAS 3938-95-2
Molecular Formula (CH3)3CCO2CH2CH3
Formula Weight 130.19
MDL Number MFCD00009129
UN Number UN3272
Beilstein 1747617
Merck Index 14,7511
Synonym Ethyl pivalate; Trimethylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1310014

  SDS
alfa aesar™
A1310014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1310036

  SDS
alfa aesar™
A1310036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1310022

  SDS
alfa aesar™
A1310022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.19
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
Quantity 25g
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Ethyl pivalate; Trimethylacetic acid ethyl ester
Flash Point 16°C (60°F)
Odor Stench
UN Number UN3272
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Melting Point −90°C
TSCA TSCA
Boiling Point 116° to 118°C
Chemical Name or Material Ethyl trimethylacetate
CAS 3938-95-2
MDL Number MFCD00009129
Merck Index 14,7511
Assay 99%
Molecular Formula (CH3)3CCO2CH2CH3
Health Hazard 1 Danger
Beilstein 1747617
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.