missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Trifluoropyruvate, TCI America™

Manufacturer:  TCI America T143425G

 View more versions of this product

Catalog No. T143425G



Specifications

Specifications

Ethyl Trifluoropyruvate
Yellow
13081-18-0
MFCD00114935
ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
CCOC(=O)C(=O)C(F)(F)F
170.087
170.09
≥98.0% (GC)
98°C
25g
C5H5F3O3
1993
KJHQVUNUOIEYSV-UHFFFAOYSA-N
ethyl 3,3,3-trifluoro-2-oxopropanoate
2737239
Clear Liquid at 20°C

Chemical Identifiers

13081-18-0
170.087
KJHQVUNUOIEYSV-UHFFFAOYSA-N
2737239
CCOC(=O)C(=O)C(F)(F)F
C5H5F3O3
MFCD00114935
ethyl trifluoropyruvate, ethyltrifluoropyruvate, ethyl 3,3,3-trifluoropyruvate, trifluoropyruvic acid ethyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, ethyl ester, ethyl 2-oxo-3,3,3-trifluoropropanoate, 3,3,3-trifluoro-2-oxopropanoic acid ethyl ester, ethyl trifluorpyruvate, ethyl-trifluoropyruvate, pubchem2013
ethyl 3,3,3-trifluoro-2-oxopropanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents